VWR® Cleanroom Tab Dividers
Catalog #: 76547-916
Supplier: VWR International
Specifications
- Description:Cleanroom tab divider
- Size:11×17"
- Cat. no.:76547-916
Specifications
About this item
11×17" cleanroom binder tab dividers.
- Latex-free
- Class 10 certified
- Supplied 5 per pack
For use with cleanroom binders. These latex-free tab dividers are class 10 certified. They are tabbed and come in sets of 5 per pack.
Customers Who Bought This Also Bought