Value Plastics Luer Fittings and Adapters
Supplier: Avantor
Certificates
About this item
Premium luer fittings for critical applications.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Manufactured from 100% virgin raw materials in a Class 100,000 environment
- Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.
Specifications
- Catalog No:
- MFLX15973-45
- MFLX15973-45BLK
- Connection type (port 1):
- Male Luer with Rotating Lock Ring
- Male Luer with Rotating Lock Ring
- Connection type (port 2):
- No. of ports:
- 2
- 2
- Warranty:
- None
- None
Specifications
Related Information
Certificates
Search by Lot NumberMFLX30800-08_345300 MFLX30800-24_332561 MFLX30800-24_NA MFLX45518-08_344366
Conformity declaration
Customers Who Bought This Also Bought