Hayward Diaphragm Valves with True Union Connections, PVC
Supplier: HAYWARD INDUSTRIAL PRODUCT INC
Certificates
About this item
Designed for use in corrosive environments.
- Multi-turn handle gives you precise control
- Position indicator shows exact location of valve
- Self draining allows for little dead volume in valve
- PVC body and EPDM diaphragm
These Hayward diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 140 °F (60 °C) and feature true-union connections.