Parker Hannifin Push-to-Connect Check Valves, Acetal
Supplier: KAMAN FLUID POWER
Certificates
About this item
Create a secure connection with your check valve.
- Constructed of durable acetal plastic
Use these push-to-connect check valves for rigid and semirigid plastic tubing. Acetal body with EPDM O-rings. Temperature range of 34 °F (1.1 °C) to 150 °F (65.6 °C).
Customers Who Bought This Also Bought