Rubberfab Nylon Clamp for Sanitary Fittings, 1" to 1-1/2"
Catalog #: MFLX12805-04
Supplier: RUBBERFAB GASKET & MOLDING
Specifications
- Connection type:Sanitary
- Description:Nylon Clamp for Sanitary Fittings, 1" to 1-1/2"
- Material:Nylon
- Warranty:1 year
- Cat. no.:MFLX12805-04
- Clamp Type:Sanitary
Specifications
About this item
Collared clamp helps prevent unintentional opening.
- All fittings are compatible with Tri-Clamp® fittings
- Excellent chemical resistance; low leachable characteristics
- Connection type:Sanitary
- Description:Nylon Clamp for Sanitary Fittings, 1" to 1-1/2"
- Material:Nylon
- Warranty:1 year
- Cat. no.:MFLX12805-04
- Clamp Type:Sanitary
Customers Who Bought This Also Bought