Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
Catalog #: MFLX30544-20
Supplier: VWR International
Specifications
- Description:Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
- Warranty:1 year
- Cat. no.:MFLX30544-20
- Fitting Material:Platinum-Cured Silicone
- Sanitary Clamp Size (Port 1):3"
- Sanitary Gasket Size:3"
Specifications
About this item
Easily sample fluid in critical high-purity system.
- Three internal ports accept thermocouple probe0
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.
- Description:Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
- Warranty:1 year
- Cat. no.:MFLX30544-20
- Fitting Material:Platinum-Cured Silicone
- Sanitary Clamp Size (Port 1):3"
- Sanitary Gasket Size:3"
Customers Who Bought This Also Bought