Granular Activated Carbon Cartridges for "Big Blue" housing
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
Certificates
About this item
Remove organic tastes/odors and chlorine.
- Constructed with granular activated carbon (GAC)
These cartridges remove organic tastes/odors and chlorine. Special design maximizes contact time and reduces channeling or bypass that is common with GAC cartridges. Included within the cartridge is a 20-micron post-filter to reduce carbon fines††.
The 20" cartridge meets NSF Standard 42 for materials.
Specifications
- Catalog No:
- MFLX01512-64
- MFLX01512-66
- Max. temperature:
- 51.7 °C (125 °F)
- 51.7 °C (125 °F)
- Warranty:
- 1 year
- 1 year