ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Supplier: BPH PUMP & EQUIPMENT INC
Certificates
About this item
Unique air valve ensures a positive shift every time – pump will not stall.
- All diaphragms, liquid chambers, and manifolds are constructed
- of 100% virgin PTFE for long life and high purity
- Unique valve requires no lubrication or prepacking.
- Easily access air valve without disassembling the pump
Pumps are manufactured under class 1000 clean room conditions to guarantee purity.
Customers Who Bought This Also Bought