Bel-Art® Two- and Three-Way Stopcocks with Hose Barb Connections
Supplier: BEL-ART PRODUCTS
Certificates
About this item
Easily adjust flow.
- Constructed of durable PVDF
- Suitable for use to a maximum temperature of 464 °F (240 °C) and maximum pressure of 5 psi
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.
Customers Who Bought This Also Bought