In-Line Flow Indicators for Liquids and Gases
Supplier: VWR International
Certificates
About this item
An inexpensive way to visually monitor flow of liquids and gases.
- Visually confirm flow with clear window
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.