Flux Pumps Sanitary Drum Pump Air Motors
Supplier: FLUX PUMPS CORP
Certificates
About this item
Configurable to your requirements.
- High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.
Specifications
- Catalog No:
- MFLX07070-70
- MFLX07070-60
- Air consumption:
- 28.9 m³/h (17 ft.³/min)
- Max. viscosity:
- 1200 cP
- 1200 cP
- Min. motor RPM:
- Motor hp:
- 1/2
- 1/2
- Power:
- Air
- Air
- Warranty:
- 6 months
- 6 months
Specifications
Customers Who Bought This Also Bought