ContactUsLinkComponent
Burkert Paddle-Wheel Sensors with Integral Fittings
Burkert Paddle-Wheel Sensors with Integral Fittings
Certificates
Burkert Paddle-Wheel Sensors with Integral Fittings
Burkert Paddle-Wheel Sensors with Integral Fittings
Certificates
Specifications
  • Max. pressure:
  • 235 psi
  • Max. operating temperature:
  • 50 °C (122 °F)
  • Repeatability:
  • ±0.4% of reading
  • Accuracy:
  • ±1% of reading
  • Warranty:
  • 1 year