VWR® Hotplates
Supplier: VWR International
Certificates
About this item
VWR® hotplates with temperature control up to 400 °C.
- White ceramic surface for excellent chemical resistance
- Digital speed control up to 400 °C
- Available with external temperature probe (displays actual sample temperature)
- 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic hotplate stirrers (heating and stirring 76549-912, 76549-918, 76549-920 and 76549-926) or magnetic stirrers (stirring only 76549-916 and 76549-924) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: CE marked.
Customers Who Bought This Also Bought