WiFi Compact Microscopes, National Optical & Scientific Instruments
Catalog #: 470337-676
Supplier: National Optical
Specifications
- Description:WIFI compact microscope
- Cat. no.:470337-676
Specifications
About this item
Ideal for hobbyists or for teaching microscopy to primary levels.
- Easy grip design
- Low-position focusing controls
- Inward facing nosepiece
- Cordless LED illumination
National is proud to introduce their own series of WiFi digital microscopes. The DCX5-205LED represents the latest in educational technology. With included Moticam X5, this WiFi digital microscope transmits live images to iOS or Android devices.
The DCX5-205LED features high-speed, full resolution imaging technology built into one of our most popular microscopes. Use it as a conventional microscope or share live images with your class using WiFi tablets or wireless laptops. High-speed, high-resolution to fit any budget.
Customers Who Bought This Also Bought