AssureWear™ Laboratory Coats, Medicom
Supplier: Medco
Certificates
About this item
Coats are made of non-woven material with front snap closure and knitted collar and cuffs.
- Non-woven material: spunbonded meltblown spunbond (SMS) polypropylene
- Knitted collar and cuffs
- Latex-free
Non-woven 3-layer SMS lab coat with knitted cuffs. Disposable SMS laboratory coats are designed to protect healthcare professionals, lab technicians, and other users from fluids and cross-contamination.
Packaging: Gowns are packaged 10/bag, 5 bags/case.
Specifications
- Catalog No:
- CA76545-380
- CA76545-382
- CA76545-376
- CA76545-378
- CA76545-372
- CA76545-374
- CA76545-370
- CA76545-386
- CA76545-384
- 76540-448
- 76540-456
- 76540-454
- 76540-460
- 76540-458
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory