SafeMask® Premier Plus™ Procedure Earloop Face Masks, Medicom
Catalog #: CA76457-714
Supplier: Medicom
Specifications
- Sterility:Non sterile
- Color:Blue
- Description:SafeMask® Premier Plus™ procedure earloop face masks
- Cat. no.:CA76457-714
Specifications
About this item
Customized comfort in a moderate-barrier mask offering ASTM Level 2 protection.
- Non-woven spunbond outer layer provides maximum protection against fluid penetration
- Shingle pleat design prevents fluid pooling and cross-contamination
- High-quality media ensures optimal filtration and breathability
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
- Made in North America
- Latex-free
ASTM Level 2 mask for procedures that produce moderate levels of spatter or spray. Adjustable nosepiece for a comfortable, personalized fit. Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation, and allow easy removal with less risk of self-contamination.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids, and particulate material transfer.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought