AssureMask® Precision™ Procedure Tie-On Face Masks, Medicom
Supplier: Medicom
Certificates
About this item
Fog-free protection with a personalized fit.
- Non-woven spun bond outer layer provides maximum protection against fluid penetration
- Ample length ties are soft, strong, ultrasonically sealed and attached to the outside of the mask to prevent pulling and irritation
- Shingle pleat design prevents fluid pooling and cross-contamination
- High-quality media ensures optimal filtration and breathability
- SureView™ Technology fog-free nose strip ensures an unobstructed view of the operative field
- Latex-free
High-quality materials and a soft inner lining enhance breathability and comfort. Smooth, ample-length ties and an adjustable nose piece provide a customized fit. Individually folded ties under each mask for easy dispensing.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids, and particulate material transfer.
Packaging: Masks are packaged 50/box, 6 boxes/case.
Customers Who Bought This Also Bought