SafeMask® Premier Elite™ Procedure Earloop Face Masks, Medicom
Supplier: Medicom
Certificates
About this item
High-barrier, ASTM Level 3 mask for higher-risk patients and procedures.
- Non-woven spun bond outer layer provides maximum protection against fluid penetration
- Shingle pleat design prevents fluid pooling and cross-contamination
- High-quality media ensures optimal filtration and breathability
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
- Made in North America
- Latex-free
Provides ASTM level 3 liquid barrier protection without compromising comfort or breathability. Soft, strong, ultrasonically sealed ear loops attached to outside of mask prevent pulling and irritation, and allow easy removal with less risk of self-contamination. Ideal for procedures with a high risk of fluid exposure.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids and particulate material transfer.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought