Specifications
- Clothing Size:XL
- Garment application:Medical
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Back fastening
- Color:Blue
- Cat. no.:CA76544-864
Specifications
About this item
Chemo-safe with patented Flexneck™ technology for fast and easy donning and removal.
- VersaGown® with Flexneck™ technology significantly reduces cross-contamination versus standard gowns
- Designed using clinician feedback and independent clinical testing
- Provides nearly two-fold reduction in contamination of hand and wrist areas
- Helps ensure instant compliance, even without training
- Innovative design allows freedom of movement without compromising protection
- Flexneck™ creates the widest neck opening on the market and has ultrasonically sealed seams to prevent tearing during donning/doffing
- Winner of the Vizient Innovative Technology Award
Patented Flexneck™ technology allows fast and easy over-the-head donning to instantly provide complete coverage and eliminate the risk posed by untied, gaping gowns that can slip and expose skin. Flexneck™ closure also ensures safe doffing by allowing gown to swiftly yet gently tear away from the body, preventing self-contamination. The AssureWear™ Chemo Tested VersaGown® is poly coated and made with thumb loop cuffs to prevent sleeves from riding up and ensure complete wrist coverage under exam gloves.
Gowns are made of non-woven material and provide full back closure. They feature flat belt, elastic cuffs with thumb loops, and over-the-head (OTH) flexible neck closure made of two elastic strings. A tape is covering all the seams.
Disposable Impervious ultrasonically sealed Isolation Gown designed to protect health professionals and patients from cross infection.