SafeMask® FreeFlow™ Procedure Earloop Face Masks, Medicom
Supplier: Medicom
Certificates
About this item
Breathe easy, see clearly, and stay comfortable with the SafeMask® FreeFlow™ Procedure Earloop Face Masks.
- Non-woven spunbond outer layer provides maximum protection against fluid penetration
- Masks feature aluminum nose- piece, flat knitted earloop, and anti-fog strip
- Omega pleats create a unique air pocket for breathability
- High-quality media ensures optimal filtration and breathability
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
- Latex-free
Innovative contoured pleats create a unique pocket that allows air to flow freely. Integrated strip prevents eyewear from fogging. Ideal for those who feel claustrophobic in ordinary masks. Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation, and allow easy removal with less risk of self-contamination.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids, and particulate material transfer.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought