Sentinel XL® OV/AG Cartridge Filters, ILC Dover
Catalog #: 76533-624
Supplier: ILC DOVER LP
Specifications
- Description:OV/AG cartridge filters
- Cat. no.:76533-624
Specifications
About this item
The OV/AG cartridge filters protect against particulates, organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, and hydrogen fluoride.
- Multiple packaging options are available
Customers Who Bought This Also Bought