Sentinel XT® Non-Incendive Blower, ILC Dover
Catalog #: 76533-600
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XT® non-incendive blower
Specifications
About this item
The Sentinel XT® Non-incendive blower, when paired with ILC Dover’s lithium polymer battery, weighs just 2.2 pounds, yet delivers 8+ CFM airflow, exceeding NIOSH requirements.
- Proven technology with 10+ year track record of reliability
- Secure lock prevents battery removal without the use of a tool
- Brushless motor design for long life and continued quiet operation
- Robust design with secondary impact protection chamber for air handling unit
This unit can be used in hazardous locations.
Customers Who Bought This Also Bought