Sentinel XL® HD BioShield Full Hood Conversion Kit - CBRN to HD Full Hood with 3 OVAG filters, ILC Dover
Catalog #: 76533-568
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XL® HD BioShield full hood conversion kit
Specifications
About this item
Convert the CBRN PAPR System into an HD system in a few easy steps.
The blower stays the same, only the hood and filters must be changed. Kit is supplied with S-2808 BioShield full hood and 3 OV/AG cartridge filters.
Customers Who Bought This Also Bought