J&W VF-23ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A high-polarity and highly substituted cyanopropyl low-bleed phase suited for the analysis of high molecular weight compounds
- High-polarity and highly substituted cyanopropyl low-bleed phase
- Engineered for accurate analysis of very polar analytes
- 100% bonded phase permits column rinsing to enhance column lifetime
- Operating temperature up to 260 °C
- Expands application ranges to higher molecular weight compounds
- Equivalent to G8 and G48
- 0.15 mm int.Ø columns available for high efficiency GC and GC/MS analyses
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SP-2330, Rtx-2330, 007-23, BPX-70, SP-2340
Agilent J&W VF-23ms is a high-polarity and highly substituted cyanopropyl low-bleed phase that enables more accurate analysis of very polar analytes, in particular, fatty acid methyl esters. Enhanced stabilization permits splitless injection, column rinsing, and use of temperatures up to 260 °C. This expands the range of possible applications compared to other 23 ms-type phases, by enabling the analysis of higher molecular weight compounds. It is equivalent to G8 and G48.
Specifications
- Catalog No:
- CAAGCP8824
- CAAGCP8822
- CAAGCP8828
- Description:
- GC Column, 7 inch cage
- GC Column, 7 inch cage
- GC Column, 7 inch cage
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- USP code:
- G8, G48
- G8, G48
- G8, G48