About this item
Optimized for FAME cis/trans separations, especially C18 isomers, even if one isomer is present at a higher concentration.
- Tuned for optimal cis/trans separation of FAMEs, especially C18 isomers
- Excellent peak shape and separation for FAME isomers, particularly if one component is present at a higher concentration
- Bonded, crosslinked, and low bleed
- High efficiency and column loadability
- Column length up to 200 m available for detailed analysis of the C18:1 isomer cluster
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W Select FAME is tuned for optimal cis/trans separations of FAMEs, especially the C18 isomers. This low-bleed, bonded column has an isothermal maximum operation temperature of 275 °C and a programmed temperature of 290 °C—a dramatic improvement of 50 °C compared to nonbonded columns. Even though this is a very polar column, its efficiency is extremely high. Columns up to 200 m are available for detailed analysis of the C18:1 isomer cluster. This column also offers three times greater loadability, further improving the shape and separation for FAME isomers, especially if one component is present at a higher concentration.
Specifications
- Catalog No:
- CAAGCP7419
- CAAGCP7420
- CAAGCP7421
- Description:
- GC Column, 7 inch cage
- GC Column, 7 inch cage
- GC Column, 7 inch cage
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G48
- G48
- G48