J&W Ultra Inert DB-BAC1 and DB-BAC2 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent DB-BAC1 Ultra Inert and DB-BAC2 Ultra Inert columns are able to provide separation of all important blood alcohol compounds.
- Superior solution for accurate blood alcohol quantification
- Optimized resolution and baseline separation of critical blood alcohol peaks
- Excellent peak shape and accurate integration of low-concentration compounds
- Accurate identification of challenging polar analytes, even at trace levels
- Each column is tested with a uniquely designed QC test mixture to verify column performance
- Rapid separation in just over 2 minutes using either t‑Butanol or n‑Propanol as an internal standard
- Agilent Blood Alcohol Checkout Mix to verify the performance of the blood alcohol workflow
Agilent J&W Ultra Inert DB-BAC1 and DB-BAC2 columns are part of the Agilent J&W Ultra Inert GC column family capable of separating t-Butanol and n-Propanol from frequently encountered interferences in blood matrix. The unique polarity of the columns provides excellent selectivity when using this column set in GC/FID/FID analysis.
Agilent ethanol calibration standards for quantitative blood alcohol analysis, traceable to both NIST and European Reference materials (ERM). Similar phases: Rtx-BAC Plus 1, Rtx-BAC Plus 2, ZB-BAC-1, and ZB-BAC-2.
Customers Who Bought This Also Bought