Specifications
- Description:CBRN Butyl hood assembly
- Size:Universal
Specifications
About this item
The only CBRN loose-fitting powered air purifying respirator (PAPR) hood designed to deliver a CAP 2 protection rating when used with the Sentinel XL® blower.
- CBRN hood requires no fit-testing, one size fits all
- Designed for use with the Sentinel XL blower
- Lay-flat design protects the hood and allows it to stow easily in a backpack
- Latex-free
The Sentinel XL® CBRN system is the only Loose-fitting Powered Air Purifying Respirator (PAPR) designed to deliver a CAP 2 protection rating. The unit protects against military warfare agents, biological threats, industrial chemical threats, and nuclear particulate threats.
The Sentinel XL® blower platform can be used to support both decon and infectious disease applications, reducing costs and simplifying protection equipment inventory. Easy to use twist and lock cartridge filters are supplied in a hermetically sealed foil pouch. NIOSH approved, filters deliver a CAP 2 protection rating.