Sentinel XL® BioShield™ Full Hood, ILC Dover
Catalog #: 76533-540
Supplier: ILC DOVER LP
Specifications
- Description:BioShield™ Full hood PAPR
- Size:Universal
- Cat. no.:76533-540
Specifications
About this item
Designed in collaboration with leading healthcare organizations, this loose-fitting PAPR with an APF of 1,000 is made from latex-free, coated spunbond polyolefin.
- Designed for use with the Sentinel XL Blower system
- 1,000 APF, offering 100x protection over N-95s
- Excellent hood stability; fit supports a range of operations
- Sewn and taped seam construction
- No fit test required
- Latex-free
This NIOSH-approved loose-fitting PAPR features an assigned protection factor of 1,000 and comes in a single-size with no fit-testing required for wearers. This loose-fitting PAPR is designed with an internal and external skirt that interfaces with most protective suits. The optical grade PET visor provides ultra-clear visibility. Cushioned head strap supports a wide range of wearers.
Certifications: NIOSH approved.
Customers Who Bought This Also Bought