Masterflex® B/T® Controller Mounting Bracket for Modular Digital Pump System
Catalog #: MFLX77111-90
Supplier: Avantor Fluid Handling
Specifications
- Description:Controller mounting bracket for B/T Modular Digital Pump System. 2 years warranty
- Cat. no.:MFLX77111-90
Specifications
About this item
Make controlling your B/T® drive easier than ever.
- Suitable for use with B/T modular digital drives
This controller mounting bracket allows a Masterflex® B/T® digital controller to be mounted directly to the pump head for ease of use.
- Description:Controller mounting bracket for B/T Modular Digital Pump System. 2 years warranty
- Cat. no.:MFLX77111-90
Customers Who Bought This Also Bought