Masterflex® Carbon Block Cartridges with Cyst Removal, Avantor®
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
Certificates
About this item
Easily remove tastes, odors, chlorine, and sediment.
- Constructed with bonded powdered activated carbon
Long-Life Carbon Block Cartridges have a polyolefin-bonded powdered activated carbon (PAC) filter. Surface area of 2.2 million sq ft. Removes tastes, odors, chlorine, and sediment. Contains approximately 12 oz of carbon. PP end caps and outer wraps. Maximum temperature is 100 °F (38 °C); maximum differential pressure is 150 psid. For optimal performance, change cartridges at 25 psid.
** Not approved for residential use in the State of California.
Specifications
- Catalog No:
- MFLX01508-97
- MFLX01508-93
- Max. temperature:
- 82,2 °C (180 °F)
- 82,2 °C (180 °F)
- Warranty:
- 1 year
- 1 year