Masterflex® Flange Kits for Mechanically Coupled Centrifugal Pumps, Avantor®
Supplier: VWR International
Certificates
About this item
Enhance the life of your pump with service flange kits.
- Constructed of stainless steel (SS) parts
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Delivery: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
Specifications
- Catalog No:
- MFLX70725-70
- MFLX70725-72
- Wetted materials:
- SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
- SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
- Warranty:
- 1 year
- 1 year