Masterflex® Remote Control Cable Connectors, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
Specifications
- Catalog No:
- MFLX07523-94
- MFLX07595-52
- MFLX07595-45
- Model:
- 07523-94
- 07595-52
- 07595-45
- Control type:
- Analog Variable Speed
- Warranty:
- 2 years