Micro Incinerators for Sterilization of Loops and Needles, Crystal
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificates
About this item
Safely sterilize metal inoculation loops and needles utilizing infrared heating in 3 to 7 seconds, while protecting the user from flames, dangerous gases and splatters.
- Two modes of operation: Stand-By and Operation
- No flames, gases or spatters
- Reliable ceramic heating element quickly reaches 900 ⁰C
- Optimum sterilization within 3 to 7 seconds
- For Model ST-14A the angle of the heating tube can be adjusted to the users preference
- Use with the Inoculation loop holder for added safety
- 2 years replacement warranty
Certifications: CE certified
Delivery: Supplied with STA01 inoculation loop holder (which allows user to safely rest loops and needles) and AC 110 V power supply.
Customers Who Bought This Also Bought