To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:SARS-CoV-2 LAMP primer mix (N/E)
- No. of Reactions:85
- Details:10X Concentration
Specifications
About this item
The SARS-CoV-2 LAMP Primer Mix (N/E) is a dual-target primer mix that recognizes the nucleocapsid (N) and envelope (E) regions of the SARS-CoV-2 genome.
- Optimized mixture of two primer sets
- Targets N and E gene regions of SARS-CoV-2 genome
- Improved detection sensitivity
Contains 2 sets of LAMP primers (F3, B3, FIP, BIP, LF, LB) and detects SARS-CoV-2 nucleic acid.
SARS-CoV-2 LAMP primer sequences (5'→ 3')
Gene E primer set sequence:
E1-F3 TGAGTACGAACTTATGTACTCAT
E1-B3 TTCAGATTTTTAACACGAGAGT
E1-FIP ACCACGAAAGCAAGAAAAAGAAGTTCGTTTCGGAAGAGACAG
E1-BIP TTGCTAGTTACACTAGCCATCCTTAGGTTTTACAAGACTCACGT
E1-LF CGCTATTAACTATTAACG E1-LB GCGCTTCGATTGTGTGCGT
Gene N2 primer set sequence:
N2-F3 ACCAGGAACTAATCAGACAAG
N2-B3 GACTTGATCTTTGAAATTTGGATCT
N2-FIP TTCCGAAGAACGCTGAAGCGGAACTGATTACAAACATTGGCC
N2-BIP CGCATTGGCATGGAAGTCACAATTTGATGGCACCTGTGTA
N2-LF GGGGGCAAATTGTGCAATTTG
N2-LB CTTCGGGAACGTGGTTGACC