Ammonium persulfate (APS) ≥98% for electrophoresis
Catalog #: IB70080
Supplier: IBI Scientific
CAS Number: 7727-54-0
Specifications
- Size:100 g
- Cat. no.:IB70080
- Supplier No.:70080
Specifications
About this item
Synonyms: Peroxidisulfuric acid diammonium salt,Ammonium persulfate,Peroxidisulphuric acid diammonium salt
:Storage temperature: Ambient
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
ADR:5.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 120 °C
:Flash Pt:
:Flash Pt:
:M.W. 228.2 g/mol
Formula:(NH₄)₂S₂O₈
MW:228.2 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003390
CAS Number:7727-54-0
EINECS:231-786-5
UN:1444
:5.1
:III
:GHS03
:GHS08
:GHS07
Customers Who Bought This Also Bought