Safety Goggles, Clear, United Scientific
Catalog #: 470332-656
Supplier: United Scientific Supplies
Specifications
- Description:Safety goggles, clear
- Cat. no.:470332-656
Specifications
About this item
Goggles feature vents to prevent fogging and an adjustable band for a comfortable fit.
- Comfortable head band
- PVC lenses
Flexible safety goggles are manufactured from PVC and are designed to protect against splashes and impact. Four vents to prevent fogging and an adjustable band for a comfortable fit.
Certifications: Safety goggles are ANSI Z87.1-2020 certified.
Customers Who Bought This Also Bought