Hand Vacuum Pump, without Gauge, United Scientific
Supplier: United Scientific Supplies
Certificates
About this item
Easy-to-use hand vacuum pump without gauge.
- Delivers approximately 15 ml per squeeze
- Removable cap and elastic valve for quick release
- Recommended for general laboratory use
This easy-to-operate hand vacuum pump delivers approximately 15 ml per squeeze to create a vacuum of 50 mm Hg, or air pressure of approximately 2 atm. Pump features a removable cap and an elastic valve to enable quick release without dismantling the pump from the line.
This product is recommended for general laboratory use, in applications that do not require high or precise vacuum conditions.
Customers Who Bought This Also Bought