Order Entry
ContactUsLinkComponent
Plastic Disposable Apron, United Scientific
Plastic Disposable Apron, United Scientific
Catalog #: 470332-710
UNITEDSCI APRON PLASTIC DISP 2MM PK20
Plastic Disposable Apron, United Scientific
Catalog #: 470332-710

Specifications

  • Description:
    Plastic disposable apron
  • Cat. no.:
    470332-710

Specifications

About this item

Disposable aprons protect clothing from splashes.

  • Constructed from polyethylene
  • 2 mm thickness
  • Pack of 20 aprons

One size fits all. Overall garment measurements are 28×46".

Customers Who Bought This Also Bought