ContactUsLinkComponent
Erlenmeyer Flask Set of 5, Borosilicate Glass, United Scientific Supplies
Erlenmeyer Flask Set of 5, Borosilicate Glass, United Scientific Supplies
Catalog #: 76531-944
FLASK GLASS ERLENMEYER SET OF 5 1000ML
Erlenmeyer Flask Set of 5, Borosilicate Glass, United Scientific Supplies
Catalog #: 76531-944
Specifications
  • Capacity:
    50 / 125 / 250 / 500 / 1000 ml
  • Description:
    Erlenmeyer flask set of 5, borosilicate glass
  • Cat. no.:
    76531-944
Specifications
About this item
Set of 5 narrow-mouth Erlenmeyer flasks.
  • Made from borosilicate glass for an excellent combination of thermal shock resistance and physical strength
  • Heavy-duty rims and durable white enamel graduations
A convenient set of five of narrow-mouth borosilicate glass Erlenmeyer flasks. Set consists of one each of 50 ml, 125 ml, 250 ml, 500 ml, and 1000 ml flasks.
Customers Who Bought This Also Bought