Specifications
- Packaging:5 vials
- Description:Linearity
- Size:3 ml
- Cat. no.:76537-088
Specifications
About this item
Linearity FD iPTH for Abbott Systems is a five level linearity set intended to simulate human patient serum samples for purpose of determining linearity, calibration verification, and verification of reportable range for iPTH.