Order Entry
ContactUsLinkComponent
Corning® Bovine calf serum, 1 L, iron-fortified, United States origin, Corning
Corning® Bovine calf serum, 1 L, iron-fortified, United States origin, Corning
Catalog #: CA76502-732
Supplier:  Corning
Corning® Bovine calf serum, 1 L, iron-fortified, United States origin, Corning
Catalog #: CA76502-732
Supplier:  Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Origin:
    United States
  • Sterility:
    Sterile
  • Description:
    Bovine calf serum
  • Size:
    1 L
  • Storage temperature:
    −40...−10°C
  • Cat. no.:
    CA76502-732

Specifications

About this item

Bovine calf serum, United States origin are collected at government approved abattoirs in the United States.

  • Cattle are less than 35 weeks of age at time of collection
  • True pooled and triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Shelf life: 60 months
  • Format: Liquid

Iron fortified to raise iron saturation to between 70 - 80% of total iron binding capacity.

Certifications: Granted a Certificate of Suitability, R0-CEP 2018-305, by the European Directorate for the Quality of Medicines (EDQM).
Customers Who Bought This Also Bought