VWR® 3D Clear Washing Buffer 10X
Catalog #: CA76463-362
Supplier: VWR International
CAS Number:
Specifications
- Description:VWR® 3D clear washing buffer 10X
- Size:200 ml
- Cat. no.:CA76463-362
Specifications
About this item
VWR® Washing Buffer, 10X, 3D Clear
- Shelf Life: 12 months
- Volume: 200 ml
Easy to Use, no special equipment required
Compatible with IF, FP and other fluorescent labels, microplate and automation compatible, reversible to follow up with 2D H&E/IHC
Storage: refrigerate after opening
Customers Who Bought This Also Bought