PCRmax® Aluminum Sealing Film, Antylia Scientific
Catalog #: 76533-888
Supplier: Argos Technologies
Specifications
- Max. I.D.:–80 - +104 °C
- Thickness:35 µm
- Cat. no.:76533-888
Specifications
About this item
Designed for storage and PCR applications – use with temperatures ranging from –80 to +104 °C.
- Exceptional seal across entire plate safeguards samples during transport or storage
- Seal on a wide range of temperatures
- Eliminates well-to-well contamination and cross-over in PCR applications
- Can be used with 96- and 384-well PCR plates
PCRmax® aluminum sealing film is designed for applications ranging from PCR and real-time PCR to ELISA and cell cultures. Aluminum material is ideal for light-sensitive samples.
Customers Who Bought This Also Bought