Reusable Maxima ESD Cleanroom Boots with Hypalon Sole, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Cleanroom boots are manufactured from 99% polyester/1% carbon fabric.
- Color-coded by size webbing strap
- Side release buckle
- Hypalon sole
Suitable for ISO Class 4 or higher cleanrooms.
Specifications
- Catalog No:
- 76515-914
- 76515-912
- 76515-910
- 76515-908
- 76515-906
- 76515-904
- 76515-902
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon