Reusable Altessa Grid Cleanroom Frocks with Snap Closure, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Cleanroom frocks are manufactured from 99% polyester/1% carbon fabric.
- Military collar
- Raglan sleeves
- ESD knit wrists
- Snap front closure
Suitable for ISO Class 5 or higher cleanrooms.
Specifications
- Catalog No:
- 76515-768
- 76515-766
- 76515-764
- 76515-762
- 76515-760
- 76515-758
- 76515-776
- 76515-774
- 76515-772
- 76515-770
- 76515-746
- 76515-790
- 76515-788
- 76515-744
- 76515-786
- 76515-742
- 76515-784
- 76515-740
- 76515-782
- 76515-738
- 76515-780
- 76515-778
- 76515-756
- 76515-754
- 76515-752
- 76515-750
- 76515-748
- Model:
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- CFRC-74
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon