VWR® Thermometer Case for Liquid-in-Glass Thermometers
Catalog #: 76491-684
Supplier: VWR International
Specifications
- Cat. no.:76491-684
- For Use With:254 mm L thermometer
Specifications
About this item
Open cases feature a screw cap with ring and small openings around bulb for circulation.
- Screw cap
- Ring for hanging
- Metal or nickel-plated brass
- Lower section surrounding the bulb with small openings for circulation
This case protects your thermometers from accidental breakage storage.
Customers Who Bought This Also Bought