Order Entry
ContactUsLinkComponent
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Catalog #: 76491-688
Supplier:  VWR International
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Catalog #: 76491-688
Supplier:  VWR International

Specifications

  • Description:
    Digital timer, single channel
  • Cat. no.:
    76491-688
  • W×H×D:
    81.3×71.1×30.5 mm
  • Accuracy:
    ±0.01% (5 s)

Specifications

About this item

Designed with 10 buttons, this timer allows direct input for faster programming of desired time.

  • Counts up and down, and displays elapsed time after countdown
  • Capacity of 99 minutes, 99 seconds
  • Audible and visual alarms
  • Last time set memory
  • Requires two AAA batteries

Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment.

Certifications: Individually tested and inspected using NIST in A2LA accredited laboratory (certification #2448.01), ensuring conformity to ISO 17025:2005. RoHS compliant.
Delivery: Supplied with flip stand, magnetic back, pocket clip, two AAA batteries, and certificate of calibration.
Customers Who Bought This Also Bought