Clear-Razeā¢ Handheld UVC Sterilization Wand, Crystal Industries
Catalog #: 76452-602
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Specifications
- Description:Clear-Razeā¢ handheld UVC sterilization wand, 5.9Ć2.0" window, 180Ā° coverage, with rechargeable battery
- Cat. no.:76452-602
- Wavelength:254 nm
- Voltage:12 V
Specifications
About this item
UVC is a known disinfectant to mitigate risks of viral or bacteriological contamination, and has been used successfully for over 40 years. With the convenience of this simple to use Clear-Razeā¢ Handheld Wand, UVC can play a vital role in a bacteria and virus elimination strategy. Proven results through third party testing.
- User safety is key; tilt sensor requires wand to be facing down to operate
- Kills 99.98% of viruses and bacteria in 10 secs, and 99.99% in 30 s
- Eliminate the use of costly and potentially hazardous chemicals
- Operates on rechargeable battery; can be used anywhere
- Low battery indicator to eliminate downtime
- 180 degrees of coverage
- For use in laboratories, hospitals, food service, janitorial, and more
The Clear-Razeā¢ UVC Handheld Wand represents a technical innovation in the evolutionary growth of UVC devices. A built-in tilt sensor protects the operator by requiring the wand to be facing downward before the UVC light is engaged. As with all UVC products, proper UVC Personal Protective Equipment should be worn.
Certifications: CE certified.
Ordering information: Supplied with UVC safety glasses and AC adapter/charger. Additional accessories available separately.
Customers Who Bought This Also Bought