Zinc Selenide Circular Optical Windows, Firebird Optics
Supplier: FIREBIRD OPTICS
Certificates
About this item
ZnSe (Zinc selenide) optical window for use in FTIR spectroscopy.
- Circular shape
- Transmission range of 1 to 14 μm
- High resistance to thermal shock and radiation damage
- Low absorption coefficient and low dispersion
Ideal for use in high power CO₂ laser systems, FTIR applications, thermal imaging and medical systems.
Surface quality: 60/40 scratch dig
Packaging: Windows are packaged in moisture-protective packaging.
Caution: Care must be taken to avoid scratching the surface as the material is soft. Not recommended for use in harsh environments, Zinc Selenide is toxic and care must be taken when handling, Always wear rubber or plastic gloves to avoid risk of contamination.
Customers Who Bought This Also Bought