About this item
An advanced, research-grade benchtop pH meter offering accurate, repeatable results.
- With selectable resolution from 0.1 to 0.001 and an intelligent i-Steward system, AB41PH is an excellent pH benchtop meter with high resolution and consistent accuracy
- Equipped with a 6.5-inch large LCD display and a touch keypad, AB41PH offers a simple and precise control experience
- The AB41PH features a 1000-item memory and password management for GLP documentation
- Provides connectivity and data output capabilities with RS232 and USB interface
i-Steward Indicators include the pH electrode condition, 'Electrode Dirty/Broken', and reminder when meter needs recalibration. Selectable resolution from 0.1 to 0.001. Perform up to a 5-point pH calibration using automatic buffer recognition of 5 predefined buffer groups. Calibration log stores ten most recent calibrations. Compact stirrer can be used to uniformly mix solutions to get fast and repeatable results at no additional cost. With multi-function touch keypad, you can operate the meter as easy as using a smartphone.
All of the important information that you may need is shown on a 6.5” large display in multiple languages. Capture your critical readings with measurement stability indicator and selectable end-points modes – Auto-Stop holds a stable reading, Continuous shows changing readings and Timed logs data at specific intervals. A 1000-item memory for measurements and calibration trail makes for efficient data documentation. Password management is suitable for GLP documentation. Standard RS232 and USB interface allows for connection to external devices such as a printer or computer for transfer or storage of data.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76518-248
- 76518-250
- mV range:
- –2000.00...+2000.00
- –2000.00...+2000.00
- mV accuracy:
- 0.3 mV
- 0.3 mV
- Temperature range:
- –10.0...125.0 °C
- –10.0...125.0 °C
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- Display:
- LCD with backlight and one-line dot matrix
- LCD with backlight and one-line dot matrix
- Connections:
- RS232, USB Device (included), GLP/GMP data output with real-time clock
- RS232, USB Device (included), GLP/GMP data output with real-time clock
- Parameters:
- pH / mV/ ORP
- pH / mV/ ORP